missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag cfDNA kit for cell-free DNA purification from plasma

Product Code. p-7159632
Click to view available options
No. of Reactions:
4 x 48 Preps
48 Preps
Unit Size:
192 reactions
48 reactions
This item is not returnable. View return policy

Product Code. 15899147

Brand: Macherey-Nagel™ 744550.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flexible magnetic bead based purification of cell-free DNA from 1–10 mL blood plasma.

NucleoMag™ DNA Plasma

  • Consistent circulating cell-free recovery from 1–10 mL plasma samples
  • Manual and automated isolation
  • Efficient purification of fragmented DNA as small as 50 bp
  • No PCR inhibition regardless of your preferred sample volume
  • Human plasma samples derived from EDTA blood draw tubes or Cell-Free DNA BCT
  • For Liquid Biopsy analysis, NGS, qPCR, ddPCR

Specifications

Capacity 0.3 μg/μL beads
Content And Storage Room temperature (18 - 25 °C)
Isolation Technology Magnetic Bead
Sample Type Human EDTA/Cell-Free DNA BCT™ Plasma
Efficiency Binding capacity: 0.3 μg/μL beads
Elution Volume Elution volume: 50 - 200 μL
For Use With (Equipment) Magnetic separator, separation plate
For Use With (Application) Isolation of cell-free DNA from plasma
Includes NucleoMag P-Beads, buffers, Liquid Proteinase K
No. of Reactions 48 Preps
Purity A260/A280: 1.6 - 1.9
Sample Volume 1 - 10 mL
Test Time 60 min/24 preps
Yield Depends on sample source, storage and quality
Form Highly Reactive Superparamagnetic Beads
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.