Learn More
Macherey-Nagel™ Nucleosil™ Quaternary Ammonium Ion Exchange HPLC Column, 5 μm, 4.6 mm x 150 mm
Column Format: Analytical Column; Carbon Load: 10%; Pore Size: 100 Å; Column Phase: Ion Exchange
Brand: Macherey-Nagel™ 720183.46
This item is not returnable.
View return policy
Specifications
Silica | |
Ammonium | |
2 mL/min | |
150 mm | |
Spherical | |
2 to 8 | |
100-5 SB | |
60°C | |
Quaternary Ammonium | |
10% | |
No | |
EC Column |
Analytical Column | |
4.6 mm | |
For HPLC, GC, TLC, SPE | |
5800 psi (400 bar) | |
5 μm | |
100 Å | |
350 m²/g | |
L14 | |
Silica | |
Ion Exchange | |
Ion Exchange | |
Analytical HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Nucleosil™ Quaternary Ammonium Ion Exchange HPLC Column, 5 μm, 4.6 mm x 150 mm > Inner Diameter: 4.6mm
Spot an opportunity for improvement?Share a Content Correction