missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSnap Plasmid Midi kit for plasmid DNA
Click to view available options
:
50 Preps
Description
NucleoSnap™ Plasmid Midi Columns
- Produces ready-to-use, transfection-grade DNA
- Isolates up to 250 μg plasmid DNA in only 35 minutes
- No alcohol precipitation necessary
- Easy procedure: DNA precipitation – filtration – washing – elution
- High-salt / ethanolic washing steps to remove contaminants
- Low-salt elution
- Typical downstream applications: transfection of common cell lines (lipofection, electroporation)
- Also suitable for standard molecular biology application such as cloning and sequencing
Specifications
Specifications
| Bacterial Endotoxins | <50 EU/μg DNA |
| Color | Clear |
| Format | Snap-off Column |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | E. coli Culture |
| Efficiency | Binding capacity: 1.5 mg |
| Elution Volume | Elution volume: 200 - 500 μL |
| Final Product Type | Transfection-Grade Plasmid DNA |
| For Use With (Equipment) | NucleoVac 24 Vacuum Manifold |
| For Use With (Application) | Isolation of plasmid DNA |
| Show More |
Safety and Handling
Product Identifier
- NucleoSnap Plasmid Midi
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : RNase; sodium hydroxide solution (diluted < 2 %), 2-propanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur