Learn More
Macherey-Nagel Bioanalysis™ NucleoSpin™ Tissue XS, Mirco Kit
XS spin kit for purification of DNA from a broad range of small samples.
Brand:  Macherey-Nagel Bioanalysis™ 740901.50
Product Code. 12728572
  • 217.00 € / 50 preps
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Two different lysis buffers plus optional addition of the chemical additive Enhancer SX allow fine tuning for maximum yield and purity. NucleoSpin™ has been designed for Gram bacteria, archaea, fungi, and algae in soil, sludge and sediment samples. NucleoSpin™ bead tubes have ceramic beads included, to be used in a bead mill or vortexer. Efficient mechanical pulping even for Gram bacteria and spores. The Inhibitor removal columns and efficient wash buffers will eliminate PCR inhibitors. DNA can mostly be used undiluted as a PCR template.
  • Highest sensitivity even for rare targets
  • Alternative lysis systems - highest yield from diverse samples
  • Ceramic beads included - most efficient cell disruption
  • Highest purity - complete removal or PCR inhibitors
Specifications
Specifications
XS Prep
50 μg (Binding Capacity)
DNA Isolation
50 Preps
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • NucleoSpin Tissue XS (50)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidium hydrochloride, polyoxyethylenesorbitan monolaurate, proteinase k