Learn More
Masterflex™ Nylon Straight Barbed Union
Product Code. 12374618
Click to view available options
For Use With:
9.52mm
12.7mm
19.05mm
25.4mm
Unit Size:
Pack of 10
Product Code. 12374618
Brand: Masterflex™ 3061234
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Diameter (Metric) Inner 25.4 mm
Material Nylon
For Use With 25.4mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 121°C
Product Type Straight Barbed Union