Learn More
Masterflex™ Nylon Y Connector
Product Code. 12314638
Click to view available options
For Use With:
3.17mm
4.76mm
6.35mm
9.52mm
Unit Size:
Pack of 10
Product Code. 12314638
Brand: Masterflex™ 3061418
Please to purchase this item. Need a web account? Register with us today!
Specifications
Diameter (Metric) Inner 9.52 mm
Material Nylon
For Use With 9.52mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 121°C
Product Type Y Connector