Learn More
Conductivity Calibration Pouches, Oakton™
Product Code. p-6827000 Shop All Cole-Parmer Products
Click to view available options
Conductivity:
702 ppm NaCl, 1000 ppm 442, 745 ppm KCl
Unit Size:
Pack of 20
Product Code. 12393780
Brand: Cole Parmer 3565311
Please to purchase this item. Need a web account? Register with us today!
Calibrate on the go with these single-use 20mL calibration pouches. Oakton™ Conductivity Calibration Pouches are sealed to ensure freshness.
  • Convenient calibration pouches are portable
  • A μS-to-ppm conversion table is printed on each pack for quick reference
  • Accuracy is ±1% at 25±C
  • Calibration pouches include a NIST-traceable calibration report supplied by the manufacture
Specifications
Chemical Name or Material Cole-Parmer Conductivity Standards
Packaging Bottle
Conductivity 702 ppm NaCl, 1000 ppm 442, 745 ppm KCl
Technique Calibration of Conductivity Instrument
Color Clear
pH 4.7 to 7.8
Boiling Point 100°C (212°F)
For Use With (Equipment) Conductivity Meter
Solubility Soluble
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 20 Single-use Pouches