Learn More
Occludin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13409-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an integral membrane protein that is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Mutations in this gene are thought to be a cause of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG), an autosomal recessive neurologic disorder that is also known as pseudo-TORCH syndrome. Alternative splicing results in multiple transcript variants. A related pseudogene is present 1.5 Mb downstream on the q arm of chromosome 5.Specifications
| Occludin | |
| Polyclonal | |
| Unconjugated | |
| OCLN | |
| occludin, OCLN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100506658, 18260, 83497 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16625, Q61146, Q6P6T5 | |
| Ocln | |
| Occludin Fusion Protein Ag4057 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |