Learn More
Diba Omnifit™ O-Rings for Chromatography Columns
Product Code. 15865564
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
3 mm
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Material:
FFKM
FKM
Unit Size:
Pack of 10
Pack of 2
Pack of 6
Product Code. 15865564
Brand: Diba 006OR10C
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Select from FKM or FFKM (Chemraz™) Omnifit™ O-Rings for Chromatography Columns
Specifications
For Use With (Equipment) Omnifit™ HiT Chromatography Columns
Quantity 6 Pk.
Diameter (Metric) Inner 10 mm
Type Liquid Chromatography Column
Length (Metric) 10 mm
Material FFKM