missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Green, 488
Shop All Bio Techne Products
Click to view available options
Quantity:
20 Assays
Unit Size:
20 assays
Specifications
Specifications
| Description | OneStep TUNEL |
| Content And Storage | Store at -20C. |
| Quantity | 20 Assays |
| For Use With (Application) | Flow Cytometry, Fluorescence Imaging |
| Product Type | Apoptosis Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction