Learn More
Thermo Scientific™ Optical Adhesive Film, for Piko PCR plates
Brand: Thermo Scientific™ ASF0020100
39.10 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
The exceptionally clear Thermo ScientificPiko PCR/qPCR Film Seal allows high transmission of fluorescence across the range of commonly-used fluorophores. When used with aPiko Thermal CyclerorPikoReal Real-Time PCR System, sample volumes as low as 5μL can be used with minimal evaporative loss. These optical adhesive films are available in the ¼-size format for Piko PCR Plates.
When used with a Piko Thermal Cycler or PikoReal Real-Time PCR System, sample volumes as low as 5 µL can be used with minimal evaporative loss. These optical films are available in the ¼-size format for Piko PCR Plates.Features of the Thermo Scientific Piko PCR/qPCR Film Seals
• Optical clarity with minimal auto-fluorescence for real-time PCR applications
• Tight sealing for sample volumes as low as 5 µL
• Adhesive backing allows easy application
• Cryo-compatible adhesive is effective to –80°C
Related Products
Optical Adhesive Film, for Piko PCR plates
Optical Heat Seal, for Piko PCR plates
Specifications
Optical | |
Film, Seal, or Applicator | |
Pressure Sensitive Optical Seal |
100 Sheets | |
Piko | |
Piko Plate |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
9Hours
21Minutes
44Seconds
25
Your input is important to us. Please complete this form to provide feedback related to the content on this product.