Learn More
Thermo Scientific™ Orion™ Conductivity Standards and Solutions
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Brand: Thermo Scientific™ 01100910
Description
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature

Specifications
Conductivity Standard | |
10 x 15 mL | |
147 μS/cm | |
4.75 to 7.75 | |
Orion pH Meter | |
Standard Solution |
Pouch | |
Yes | |
Clear | |
100°C | |
Soluble | |
Conductivity Standard |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.