Learn More
Thermo Scientific™ Orion™ Star A Series Electrode Holder for Electrode Stands
Use the Thermo Scientific™ Orion™ Star A Series Electrode Holder with meter-attached or swing arm electrode stand to organize sensors for easier movement, rinsing and storage
Brand:  Thermo Scientific™ STARA-ELHD
80.18 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 16286581
  • 84.40 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use the Thermo Scientific™ Orion™ Star A Series Electrode Holder with a meter-attached or swing arm electrode stand to organize your sensors for easier movement in and out of samples and simplified rinsing and storage. The electrode holder accepts up to six sensors using 12mm holder with optional ATC probe insert, 20° angle holder for gas sensing ISEs, 15mm holder for conductivity and DO probes, 12mm flex clip holder for pH, ORP and ion selective electrodes, center stirrer probe holder and removable RDO probe holder.
The Orion Star A and Versa Star electrode holder head has identifiable slots and offers the following benefits:
  • Adjustable probe heights reduce the amount of sample needed for testing 
  • Easy removal from clips enables easier electrode maintenance and saves time
  • The 20° angle position for gas-sensing ISEs makes it easier to keep bubbles off the sensor for accurate readings
  • Dedicated stirrer port for Orion Star stirrer probe aids in quick and easy sample mixing
  • Fixed front slot works with ATC probes (using the included adapter) or other 12mm dia probes
Use to update existing swing arm stands (090043) or as a replacement electrode head on the Orion Star A and Versa Star electrode arm and holder (STARA-BEA).
Includes:
Redesigned electrode holder head, ATC probe adapter and RDO probe clip
Requires:
Electrode arm and base (attached to meter or separate)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orion Electrode Holder Head with ATC probe adapter and RDO probe clip
Electrode Stand/Holder
Redesigned electrode holder for up to six electrodes and sensors (purchase stand and base separately)
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Orion™ Star A Series Electrode Holder for Electrode Stands >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.