missing translation for 'onlineSavingsMsg'
Learn More

OTS964 hydrochloride, MedChemExpress

Product Code. 16485876
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485876

Brand: Medchem Express HY12467

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OTS964 hydrochloride is an orally active, high affinity and selective TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor with an IC50 of 28 nM. OTS964 hydrochloride is also a potent inhibitor of the cyclin-dependent kinase CDK11, which binds to CDK11B with a Kd of 40 nM.

OTS964 hydrochloride is an orally active, high affinity and selective TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor with an IC50 of 28 nM. OTS964 hydrochloride is also a potent inhibitor of the cyclin-dependent kinase CDK11, which binds to CDK11B with a Kd of 40 nM.

Specifications

Chemical Name or Material OTS964 hydrochloride
CAS 1338545-07-5
Color White
Purity Grade Notes Research
Molecular Formula C23H25ClN2O2S
Quantity 100 mg
Solubility Information DMSO : ≥ 83.33 mg/mL (194.26 mM) ∣H2O : 1.43 mg/mL (3.33 mM; Need ultrasonic)
SMILES OC1=CC(C)=C2C(C3=C(SC=C3)C(N2)=O)=C1C4=CC=C([C@@H](C)CN(C)C)C=C4.Cl
Molecular Weight (g/mol) 428.97
Formula Weight 428.97
Percent Purity 95.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.