Learn More
Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts
Click to view available options
Type:
End Gates
Gasket
Gel Tray
Unit Size:
2 sets
Each
Description
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned so you can arrange up to 10 sample sets on the same gel
- Unique electrode placement allows this gel to be run quickly without compromising the resolution
Specifications
Specifications
| For Use With (Equipment) | Owl A1 Large Gel Electrophoresis System |
| Product Line | Owl |
| Type | Gasket |
| Includes | Gasket Pair |