missing translation for 'onlineSavingsMsg'
Learn More

OXA (17-33), Tocris Bioscience™

Product Code. 30002008
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective peptide OX1 agonist

Potent and selective peptide orexin OX1 receptor agonist (EC50 values are 8.29 and 187 nM for OX1 and OX2 receptors respectively). Truncated form of orexin A (Cat. No. 1455).

Specifications

CAS 343268-91-7
Molecular Formula C79H125N23O22
Quantity 1 mg
Synonym orexin a 17-33
InChI Key JPRBZUXULYLVPJ-HYNYKCRDSA-N
SMILES CCC(C)C(C(=O)NC(CC(C)C)C(=O)NC(C(C)O)C(=O)NC(CC(C)C)C(=O)N)NC(=O)CNC(=O)C(C)NC(=O)C(C)NC(=O)C(CC1=CN=CN1)NC(=O)C(CC(=O)N)NC(=O)CNC(=O)C(C)NC(=O)CNC(=O)C(CC2=CN=CN2)NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)C(CCC(=O)O)NC(=O)C(CC3=CC=C(C=C3)O)N
Molecular Weight (g/mol) 1749.008
PubChem CID 77846288
Formula Weight 1749
Chemical Name or Material OXA (17-33)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.