missing translation for 'onlineSavingsMsg'
Learn More

Oxaliplatin, Tocris Bioscience™

Product Code. 30001455
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

DNA cross-linking antitumor agent

Antitumor agent that forms platinum-DNA adducts. Causes intra- and interstrand DNA crosslinks blocking DNA replication and transcription. Displays higher cytotoxicity and lower nephrotoxicity than analog cisplatin (Cat. No. 2251) and shows antitumor activity in cell lines with acquired cisplatin resistance.

Specifications

CAS 61825-94-3
Molecular Formula C8H14N2O4Pt
MDL Number MFCD00866327
Quantity 50 mg
Synonym Oxalato[(1R-trans)-1, 2-cyclohexanediamine]platinum(II)
InChI Key ZROHGHOFXNOHSO-WHMNLBETNA-L
SMILES O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
Molecular Weight (g/mol) 397.29
Formula Weight 397.29
Percent Purity >99%
Chemical Name or Material Oxaliplatin
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'