Learn More
Thermo Scientific™ Oxoid™ Clostridium difficile Test Kit
Thermo Scientific™ Oxoid Clostridium difficile Test Kit is a rapid latex agglutination test to identify C. difficile from culture media.
Brand: Thermo Scientific™ DR1107A
Description
Thermo Scientific™ Oxoid Clostridium difficile Test Kit is a rapid latex agglutination test used to identify Clostridium difficile from selective culture media.
Specifications
50 Tests/Kit |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4375150 | Certificate of Analysis | 28/02/2023 | DR1107A |
4375150 | Certificate of Analysis | 30/07/2022 | DR1107A |
4322907 | Certificate of Analysis | 13/07/2021 | DR1107A |
4319890 | Certificate of Analysis | 19/06/2021 | DR1107A |
4316042 | Certificate of Analysis | 25/05/2021 | DR1107A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.