missing translation for 'onlineSavingsMsg'
Learn More

p57Kip2Ab-6 Mouse Monoclonal Antibody, Epredia™

Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-6, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1062-R7

391.60 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 12628786

  • 457.00 € / 7mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.

Host Species: Mouse

Clone: 57P06; same as KP10

Isotype: IgG2b/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: Not determined

Immunogen: Recombinant human p57Kip2 protein

Molecular Weight: 57kDa

Positive Control: LS174T cells, Colon CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native verified)
Specifications

Specifications

p57superscriptKip2superscript Ab-6
Monoclonal
Unconjugated
Recombinant human p57superscriptKip2superscript protein
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin), Immunoprecipitation
57P06; same as KP10
Mouse
7 mL
Cell Cycle
Human, Mouse
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.