missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PADI4 Recombinant Protein

335.00 € - 508.00 €

Specifications

Accession Number AAH25718.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 23569
Molecular Weight (g/mol) 98.67
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16137482
View Documents Promotion Details
Abnova™
H00023569-P01-10
10 ug
335.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
16147482
View Documents Promotion Details
Abnova™
H00023569-P01-25
25 ug
508.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH25718.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
98.67
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PADI4
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
23569
PADI4 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQRTLIRVTPEQPTHAVCVLGTLTQLDICSSAPEDCTSFSINASPGVVVDIAHSPPAKKKSTGSSTWPLDPGVEVTLTMKAASGSTGDQKVQISYYGPKTPPVKALLYLTAVEISLCADITRTGKVKPTRAVKDQRTWTWGPCGQGAILLVNCDRDNLESSAMDCEDDEVLDSEDLQDMSLMTLSTKTPKDFFTNHTLVLHVARSEMDKVRVFQATRGKLSSKCSVVLGPKWPSHYLMVPGGKHNMDFYVEALAFPDTDFPGLITLTISLLDTSNLELPEAVVFQDSVVFRVAPWIMTPNTQPPQEVYACSIFENEDFLKSVTTLAMKAKCKLTICPEEENMDDQWMQDEMEIGYIQAPHKTLPVVFDSPRNRGLKEFPIKRVMGPDFGYVTRGPQTGGISGLDSFGNLEVSPPVTVRGKEYPLGRILFGDSCYPSNDSRQMHQALQDFLSAQQVQAPVKLYSDWLSVGHVDEFLSFVPAPDRKGFRLLLASPRSCYKLFQEQQNEGHGEALLFEGIKKKKQQKIKNILSNKTLREHNSFVERCIDWNRELLKRELGLAESDIIDIPQLFKLKEFSKAEAFFPNMVNMLVLGKHLGIPKPFGPVINGRCCLEEKVCSLLEPLGLQCTFINDFFTYHIRHGEVHCGTNVRRKPFSFKWWNMVP
PAD/PAD4/PADI5/PDI4/PDI5
PADI4
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications