Learn More
Mettler Toledo™ Parking Beaker for EasyPlace Electrode Arm
Product Code. 17551016 Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
EasyPlace
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17551016
Brand: Mettler Toledo™ 30671571
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Parking beaker for the EasyPlace electrode arm. Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements, or use it to store the wetting cap while measuring.
  • Safe Short-Term Storage
  • The parking beaker (with insert) stores a sensor in a suitable storage solution without removing it from the EasyPlace - never store your sensor dry
TRUSTED_SUSTAINABILITY
Specifications
Type Parking Beaker, EasyPlace
For Use With (Equipment) EasyPlace