Learn More
Mettler Toledo™ Parking Beaker for EasyPlace Electrode Arm
Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
EasyPlace
Unit Size:
Each
Description
- Safe Short-Term Storage
- The parking beaker (with insert) stores a sensor in a suitable storage solution without removing it from the EasyPlace - never store your sensor dry
Specifications
Specifications
| Type | Parking Beaker, EasyPlace |
| For Use With (Equipment) | EasyPlace |