Learn More
Carl Roth™ PC Cryo Box
Brand:  Carl Roth™ K594.1
Encompass
Product Code. 10188951
  • 20.85 € / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Slip Lid
Polycarbonate
Cryo Box
121°C
1 Each
133 x 133 mm
133 mm
52 mm
Green
Square
100 (1 to 2 mL)
-196°C
-196°C to +121°C
133 x 133 x 52 mm
133 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Carl Roth™ PC Cryo Box