Learn More
Greiner Bio-One PCR Strip Tubes without Cap
Product Code. 10787672
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Unit Size:
Pack of 125
Pack of 1250
Product Code. 10787672
Brand: Greiner Bio-One 673210
Please to purchase this item. Need a web account? Register with us today!
Edge
For use in research. Greiner Bio-One PCR Strip Tubes without Cap are free of detectable DNase, Rnase and human DNA.
  • Non-Pyrogenic
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Color Natural
Quantity 1250
Material Polypropylene
Autoclavable Not Autoclavable