missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PDE9A 293T Cell Transient Overexpression Lysate (Denatured) (T02)

Human overexpression lysate

Brand:  Abnova™ H00005152-T02.100uL


Product Code. 16102545

  • 209.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

293T
PDE9A 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 61.7kDa
Transient overexpression cell lysate was tested with anti-PDE9A antibody (Mfr. No. H00005152-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_001001567.1
pCMV-PDE9A full-length
100 μL
Human
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL PDE9A 293T Cell Transient Overexpression Lys

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Promotions

Promotions