missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (MM0004-8A89) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60969-0.025MG
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PDGFR alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100-400, Immunohistochemistry-Paraffin 1:100-1:400 | |
| P16234 | |
| PDGFRA | |
| N-terminal recombinant human platelet derived growth factor receptor alpha fragment. | |
| 0.025 mg | |
| Protein Kinase | |
| 5156 | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| MM0004-8A89 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| PDGF Receptor alpha. No cross-reactivity is shown with hPDGF Receptor beta. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction