missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK4 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-07049H
This item is not returnable.
View return policy
Description
PDK4 Polyclonal specifically detects PDK4 in Human samples. It is validated for Western Blot.
Specifications
| PDK4 | |
| Polyclonal | |
| Western Blot | |
| Q16654 | |
| PDK4 | |
| Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 200-250). [Swiss-Prot# Q16654] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 94% homology with rat and 88% homology with mouse. | |
| Store at 4C in the dark. |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial | |
| Rabbit | |
| 49 kDa | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction | |
| 5166 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction