missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PDLIM3 Recombinant Protein
335.00 € - 508.00 €
Specifications
Accession Number | AAH27870 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 27295 |
Molecular Weight (g/mol) | 60.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128752
|
Abnova™
H00027295-P01.10ug |
10 ug |
335.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16138752
|
Abnova™
H00027295-P01.25ug |
25 ug |
508.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH27870 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
60.5 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PDLIM3 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
27295 | |
PDLIM3 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPQTVILPGPAPWGFRLSGGIDFNQPLVITRITPGSKAAAANLCPGDVILAIDGFGTESMTHADAQDRIKAAAHQLCLKIDRGETHLWSPQVSEDGKAHPFKINLESEPQEFKPIGTAHNRRAQPFVAAANIDDKRQVVSASYNSPIGLYSTSNIQDALHGQLRGLIPSSPQNEPTASVPPESDVYRMLHDNRNEPTQPRQSGSFRVLQGMVDDGSDDRPAGTRSVRAPVTKVHGGSGGAQRMPLCDKCGSGIVGAVVKARDKYRHPECFVCADCNLNLKQKGYFFIEGELYCETHARARTKPPEGYDTVTLYPKA | |
ALP/DKFZp686L0362 | |
PDLIM3 | |
Wheat Germ (in vitro) | |
GST |