Learn More
Karl Hecht™ Pear Shaped Flasks
Volume indicated by durably marked label on flask.
Brand: Karl Hecht™ 41920044
This item is not returnable.
View return policy
Description
- Round bottom
- With standard ground, joint size 14/23
- Manufactured from borosilicate glass 3.3
Specifications
Clear | |
Borosilicate Glass (3.3) | |
14/23 |
50 mL | |
Pear | |
Round |