Learn More
Thermo Scientific™ PEEK Capillary Tubing
Product Code. 10609993 Shop All Thermo Scientific Products
Click to view available options
Overall Length (Metric):
5 cm
10 cm
20 cm
30 cm
50 cm
100 cm
Diameter (English) Inner:
0.003 in.
0.005 in.
0.007 in.
0.010 in.
0.020 in.
Color:
Blue
Natural
Orange
Red
Yellow
Unit Size:
Each
Product Code. 10609993
Brand: Thermo Scientific™ 3700520
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors.
Use Thermo Scientific™ Rheodyne™ Sample Loops for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
  • Broad chemical compatibility
  • Biocompatible
  • Easily cut to desired length
  • Appropriate for many HPLC applications
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
TRUSTED_SUSTAINABILITY
Specifications
For Use With Rheodyne HPLC Sample Injectors
Type Capillary Tubing
Color Red
Diameter (English) Inner 0.005 in.
Diameter (Metric) Inner 0.127 mm
Overall Length (English) 7.874 in.
Overall Length (Metric) 20 cm
Material PEEK
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.