missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bode Chemie™ Peha-Soft™ Nitrile Fino Powder-Free Glove
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Latex-free disposable examination and protective gloves made from particularly soft, purple-colored nitrile rubber, powder-free
- Easy to put on thanks to the inner coating; textured finger area for optimal grip
- Highly elastic, particularly tear-resistant and safe; excellent fit and excellent tactile sensitivity
- To wear left and right.
Specifications
Specifications
Certifications/Compliance | EN 374, Cat. III |
Applications | Laboratory |
Color | Purple |
Disposable | Single-use |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Product Type | Nitrile Examination Glove |
Size | X-Small |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'