missing translation for 'onlineSavingsMsg'
Learn More

Hartmann Paul™ Peha-Soft™ Nitrile Guard Powder-Free Glove

Product Code. 15369857
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Peha-Soft™ latex-free examination and protective glove made of synthetic nitrile rubber with the long cuff.

  • Latex-free disposable examination and protective gloves made of soft, blue-colored nitrile rubber, powder-free
  • Extra long cuff for optimal protection
  • Easy to put on thanks to chlorinated inner coating
  • Textured finger area for optimal grip
  • Highly elastic and particularly tear-resistant
  • Excellent fit and excellent tactile sensitivity
  • To wear left and right

Specifications

Certifications/Compliance EN 455 and EN 374
Chlorinated Yes
Applications Laboratory
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.