missing translation for 'onlineSavingsMsg'
Learn More

Bode Chemie™ Peha-Soft™ Nitrile White Powder-Free Glove

Product Code. 15728478
Click to view available options
Size:
X-Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Peha-soft™ latex-free examination and protective glove made of white, synthetic nitrile rubber.

  • Latex-free disposable examination and protective gloves made of soft nitrile rubber, powder-free
  • Easy to put on thanks to the chlorinated inner coating
  • Textured finger area for optimal grip
  • Highly elastic and particularly tear-resistant
  • Excellent fit and excellent tactile sensitivity
  • To wear left and right.

Specifications

Certifications/Compliance EN 455 and EN 374
Chlorinated Yes
Applications Healthcare, Laboratory
Color White
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'