missing translation for 'onlineSavingsMsg'
Learn More

Pemetrexed, Tocris Bioscience™

Product Code. 30000991
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity dihydrofolate reductase inhibitor; also inhibits thymidylate synthase, GARFT and AICART

High affinity dihydrofolate reductase inhibitor (Ki = 7 nM). Also inhibits thymidylate synthase, AICART and glycinamide ribonucleotide formyltransferase (Ki values are 109 nM, and 3.5 μM and 9.3 μM, respectively). Indirectly activates AMPK. Inhibits proliferation of cancer cell lines in vitro.

Specifications

CAS 137281-23-3
Molecular Formula C20H21N5O6
Linear Formula C20H21N5O6
Quantity 50 mg
Synonym Pemetrexed
Solubility Information Soluble to 100mM in DMSO
InChI Key WBXPDJSOTKVWSJ-ZDUSSCGKSA-N
SMILES C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O
IUPAC Name (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid
Molecular Weight (g/mol) 427.417
PubChem CID 446556
ChEBI CHEBI:63616
Formula Weight 427.41
Percent Purity >98%
Physical Form Solid
Chemical Name or Material N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.