missing translation for 'onlineSavingsMsg'
Learn More
Petroleum ether, pure, boiling range ca. 180-280°C, Thermo Scientific Chemicals Product Code.: 10260513

Petroleum ether, pure, boiling range ca. 180-280°C, Thermo Scientific Chemicals

Product Code. 10260513
25 L, Metal drum
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10260513

Brand: Thermo Scientific Acros 387490250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561

Specifications

Color Colorless
Density 0.7900g/mL
Boiling Point 200.0°C to 250.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Packaging Metal drum
Refractive Index 1.4300 to 1.4500
Quantity 25 L
Merck Index 15,5342
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Grade Pure
Physical Form Liquid
Chemical Name or Material Petroleum ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466649 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number387490050,387490250,387490010,387490025
Lot NumberA0461968 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog Number387490050,387490250,387490010,387490025
Lot NumberA0461106 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number387490050,387490250,387490010,387490025
Lot NumberA0459899 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number387490050,387490250,387490010,387490025
Lot NumberA0451877 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number387490050,387490250,387490010,387490025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range ca. 180-280°C
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Petroleum ether, pure, boiling range ca. 180-280°C, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.