Learn More
Mettler Toledo™ pH Combination Electrodes
Brand:  Mettler Toledo™ 59903231
 View more versions of this product
Product Code. 10094523
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
pH Combination Electrode
0 to 12
Use in reactors, tanks, open basins and transfer lines under harsh chemical process conditions.
200 mm
0°C to 100°C