Learn More
Magle Life Sciences™ Phadebas™ Saliva Amylase-detection Forensic Paper
Used for presumptive testing of suspicious stains. Phadebas™ Saliva Amylase-detection Forensic Paper is based on the Phadebas Amylase Test tablets but follows a separate method developed by the Forensic Biology labs.
Brand:  Magle Life Sciences™ 1317
 View more versions of this product
Product Code. 12825562
  • 1195.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Pre-manufactured papers;
  • Are applicable directly from package;
  • Include Batch number and Expiry date;
  • Are easy to handle and use;
  • Have negligible batch variations;
  • Give no difference between labs and consequently have higher credibility as evidence material
Specifications
Specifications
395mm x 15m roll
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Magle Life Sciences™ Phadebas™ Saliva Amylase-detection Forensic Paper > Size: 395mm x 15m roll