Learn More
Bochem™ Pharma scoop
Bochem™ Pharma scoop is a highly polished scoop with a handle.
Brand:  Bochem™ 8860
Product Code. 15780757
  • 152.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Polished
  • Includes handle
Specifications
Specifications
Stainless Steel (316)
100 mL
100 mm
1.22mm wall thickness
180 mm
50 mm