Learn More
Kirsch Philipp™ Pharmaceutical Refrigerator
Storage for sensitive chilled goods.
Brand:  Kirsch Philipp™ 10589
Product Code. 10630615
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: Ten height adjustable drawers
This item is not returnable. View return policy
Description
Description
  • Individual drawer division with adjustable length and cross dividers
  • Recirculating air-cooling
  • Optimized ventilation plates to minimize interior temperature differences
  • Automatic defrosting
  • Melt water evaporation
  • Easy-to-read LED temperature display
  • Easy accessible control panel with key switch
  • Micro-processor controlled temperature
  • Thawing regulation to maximize temperature stability
  • Minimum/Maximum temperature memory
  • Safety control to prevent freezing
  • Optical and acoustical alarm including power failure alarm
  • Potential free contact for alarm transmission
  • RS-485 interface
  • Low-noise compressor
Specifications
Specifications
460 L
24 kg
57 x 60 x 128 cm
220/240 V
143 kg (gross)
2°C to 20°C
Storage of pharmaceutical products
140 kw/24 hours
50 - 60 Hz 180 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kirsch Philipp™ Pharmaceutical Refrigerator > Pharmaceutical Refrigerator