Learn More
Medicago Phosphate Buffered Saline Tablets, pH 7.4
Specifically developed for immunological and microbiological laboratories. Medicago Phosphate Buffered Saline Tablets, pH 7.4 are provided as pre-weighed tablets with a final volume of 500 or 1000ml per tablet.
Brand:  Medicago 09-9400-100
 View more versions of this product
Product Code. 11330439
  • 423.00 € / Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Among biological buffers, PBS is one of the most commonly used. The buffer is isotonic and non-toxic to cells and has the ability to maintain their osmolarity. The buffer is suitable for washing procedures in cell cultures and for immunoassays such as ELISA and immuno-histochemical procedures. It is often used for sample dilution in molecular biology and as protein diluent in Western blotting. Furthermore, the buffer can function as an equilibrator for gel filter columns.
  • Formulated from analytical grade reagents
  • Autoclavable
  • Avaliable as convenient tablets that are ready to use in minutes
Specifications
Specifications
White
Analytical Grade
Immunoassays, tissue and cell culture procedures, immuno-histochemical procedures, microbiological procedures, and sample dilution
Tablet
25°C
100 x 1000 mL tablets
7.4