missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Iodination Reagent (1,3,4,6-tetrachloro-3α,6α-diphenyl-glycoluril)

Activate iodine for incorporation of iodine-125 labels into protein tyrosyl groups using this tetrachloro-diphenyl-glycoluril oxidizing agent.

Brand:  Thermo Scientific™ 28600

Product Code. 10404414

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Iodination Reagent is 1,3,4,6-tetrachloro-3a,6a-diphenyl-glycoluril, an oxidizing agent that efficiently activates iodine for spontaneous incorporation into protein tyrosyl groups.

Unlike chloramine-T, Pierce Iodination Reagent can be coated on glassware to enable solid-phase iodine-labeling. As a result, reducing agents are not necessary in the procedure.

Highlights:

  • Extremely stable and virtually insoluble in water
  • Solubility in chloroform permits plating aliquots of the reagent on the walls of the glass or plastic iodination vessels; plated vessels can be prepared in advance and stored in a dessicator
  • Allows rapid iodination of proteins in an aqueous reaction mixture containing radioactive Na125I
  • Side reactions are negligible
  • No reducing reagent is required because reactions are terminated by simply decanting the reaction solution away from the plated iodination reagent
  • Specific activities of 10,000cpm of 125I per microgram of protein and labeling efficiencies of 60% are easily achieved
  • Can be used to iodinate phenolic groups on crosslinkers or other protein modification reagents either before or after a protein, peptide or cell surface is reacted with the modification reagent
  • Small amounts of protein can be iodinated
  • More efficient than Chloramine-T or Lactoperoxidase
  • Iodinate phenolic groups on crosslinking reagents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Iodination Reagent
Upon receipt store refrigerated and desiccated. Product shipped at ambient temperature.
Iodine-125
1 g
Pierce™
Radioisotope
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberXI324298 Certificate TypeCertificate of Analysis Date10/09/2022 Catalog Number28600
Lot NumberWF319559 Certificate TypeCertificate of Analysis Date02/07/2021 Catalog Number28600
Lot NumberVL308627 Certificate TypeCertificate of Analysis Date23/12/2020 Catalog Number28600
Lot NumberVJ309178 Certificate TypeCertificate of Analysis Date04/11/2020 Catalog Number28600
Lot NumberVH311208 Certificate TypeCertificate of Analysis Date03/09/2020 Catalog Number28600
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Iodination Reagent (1,3,4,6-tetrachloro-3α,6α-diphenyl-glycoluril) > Iodination reagent powder; 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.