missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Pinacidil Monohydrate K+ channel activator
K+ channel activator
Brand: Novus Biologicals™ NBP2-62756-50MG
This item is not returnable.
View return policy
Description
PubChem ID: 55329
Application Notes: May be dissolved in DMSO (25 mg/mL, warm); or ethanol (25 mg/mL, warm)

Specifications
85371-64-8 | |
C13H19N5· H2O | |
50 mg | |
Pinacidil Monohydrate K+ channel activator |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
>98% (HPLC); NMR (conforms) | |
Synthetic | |
Powder |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Pinacidil Monohydrate K+ channel activator > 50 mg
Spot an opportunity for improvement?Share a Content Correction