Learn More
Pinacolone, 97%, Thermo Scientific Chemicals
85.90 € - 331.00 €
Chemical Identifiers
CAS | 75-97-8 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.161 |
MDL Number | MFCD00008846 |
InChI Key | PJGSXYOJTGTZAV-UHFFFAOYSA-N |
Synonym | pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone |
PubChem CID | 6416 |
IUPAC Name | 3,3-dimethylbutan-2-one |
SMILES | CC(=O)C(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474023
|
Thermo Scientific Alfa Aesar
A10289.22 |
100 g |
85.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484023
|
Thermo Scientific Alfa Aesar
A10289.36 |
500 g |
331.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.
Solubility
Miscible with water, chloroform, ethanol, ether, acetone and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
75-97-8 | |
100.161 | |
PJGSXYOJTGTZAV-UHFFFAOYSA-N | |
6416 | |
CC(=O)C(C)(C)C |
C6H12O | |
MFCD00008846 | |
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone | |
3,3-dimethylbutan-2-one |
Specifications
75-97-8 | |
0.803 g/mL | |
5°C (41°F) | |
C6H12O | |
1.397 | |
UN1224 | |
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone | |
PJGSXYOJTGTZAV-UHFFFAOYSA-N | |
3,3-dimethylbutan-2-one | |
6416 | |
97% |
°C to 53°C | |
105°C to 106°C | |
97% | |
(CH3)3CC(O)CH3 | |
MFCD00008846 | |
14,7440 | |
Miscible with water,chloroform,ethanol,ether,acetone and carbon tetrachloride. | |
CC(=O)C(C)(C)C | |
100.161 | |
100.16 | |
Pinacolone |
Safety and Handling
GHS H Statement
H225-H302
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H225-H302
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 200-920-4
missing translation for 'rtecsNumber' : EL7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only