Learn More
Brady™ Pipe Banding Tape
Click to view available options
Dimensions (LxW):
33 m x 100 mm
33 m x 25 mm
33 m x 50 mm
Color:
Black
Blue
Brown
Gray
Green
Orange
Purple
Red
White
Yellow
Unit Size:
1 roll
Description
- Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
Specifications
Specifications
| Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, CSA Accepted, UL Recognized, |
| Description | Pipe Banding Tape |
| Adhesive Type | Permanent Acrylic, Permanent |
| Color | Purple |
| Finish | Satin |
| Format | Tape |
| Material | Polyester |
| Product Type | Pipe Marker |
| For Use With (Application) | Pipe and valve marking |
| No. per Pack | 1 Roll |
| Show More |