Learn More
Brady™ Pipe Banding Tape
Product Code. 12759884
Click to view available options
Dimensions (LxW):
33 m x 100 mm
33 m x 25 mm
33 m x 50 mm
Color:
Black
Blue
Brown
Gray
Green
Orange
Purple
Red
White
Yellow
Unit Size:
1 roll
Product Code. 12759884
Brand: Brady™ 275200
Only null left
Add to basket
Add to basket
Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
  • Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, CSA Accepted, UL Recognized,
Description Pipe Banding Tape
Adhesive Type Permanent Acrylic, Permanent
Color Yellow
Finish Satin
Format Tape
Material Polyester
Print Technology Thermal Transfer
Product Type Pipe Marker
For Use With (Application) Pipe and valve marking
No. per Pack 1 Roll
Quantity 1 Ea.
Temperature Range (English) -112°F to +230°F
Temperature Range (Metric) -80°C to +110°C
Thickness (English) 0.0028 in.
Thickness (Metric) 0.071 mm
Dimensions (LxW) 33 m x 25 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 0.98 in.
Width (Metric) 25 mm
Alternate Part Number PIPEBANDINGTAPEYELLOW25MMX33M