missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperacillin Sodium Salt, MP Biomedicals™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
Type | Piperacillin Sodium Salt |
Physical Form | Crystalline Powder |
IUPAC Name | sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
Formula Weight | 539.5 |
Color | White |
Quantity | 10 g |
Chemical Name or Material | Piperacillin Sodium Salt |
Safety and Handling
Product Identifier
- Piperacillin Sodium Salt
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P321-Specific treatment (see on this label).
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Piperacillin SodiumSalt
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'