Learn More
Greiner Bio-One Pipette Tips, 200-1000 μL, Conductive
Product Code. 15965351
Click to view available options
Volume (Metric):
200 to 1000 μL
Product Code. 15965351
Brand: Greiner Bio-One 686296
Only null left
Add to basket
Add to basket
Specifically designed for use on robotic systems with automated liquid level sensors.
  • For robotic systems with automated liquid level sensors.
  • The graphite incorporated into the polypropylene allows the tip to become an integral part of the electrical resistance sensors.
  • Reproducibility, accuracy and minimum carry-over are optimized to highly critical levels.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 to 1000 μL
Volume (Metric) Dispensing 200 to 1000 μL
Sterility Non-sterile
Color Black
Tip Style Conductive
Packaging Bulk
Quantity 250/Pk, 5000/Cs
Material Polypropylene