Learn More
Greiner Bio-One Pipette Tips, 200-1000 μL, Conductive
Click to view available options
Volume (Metric):
200 to 1000 μL
Description
- For robotic systems with automated liquid level sensors.
- The graphite incorporated into the polypropylene allows the tip to become an integral part of the electrical resistance sensors.
- Reproducibility, accuracy and minimum carry-over are optimized to highly critical levels.
Specifications
Specifications
| Volume (Metric) | 200 to 1000 μL |
| Volume (Metric) Dispensing | 200 to 1000 μL |
| Sterility | Non-sterile |
| Color | Black |
| Tip Style | Conductive |
| Packaging | Bulk |
| Quantity | 250/Pk, 5000/Cs |
| Material | Polypropylene |