Learn More
Greiner Bio-One Pipette Tips, 200-1000 μL, Universal, Blue
Product Code. 10194621
Click to view available options
Packaging:
Bag
Easyload Rack
Quantity:
250/Bag, 5000/Box
60/Rack, 2400/Box
Unit Size:
Pack of 1000
Pack of 2400
This item is not returnable. View return policy
Product Code. 10194621
Brand: Greiner Bio-One 740265
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Manufactured from high-grade polypropylene for improved performance, fit, and stability
  • Manufactured from high-grade polypropylene for optimum performance and fit
  • Polypropylene has a high breaking strength, is dimensionally stable, heat-resistant up to approx. 40°C, i.e. is autoclavable, and cold-resistant down to −190°C
  • Free of heavy metals
  • Manufactured under production conditions certified as conforming to the ISO 9001 standard.
  • Tested for conformance with the international standard DIN ISO 8655
  • Recommended when pipettes from different manufacturers are used at the same time
  • Ideally suited for Brand™; Eppendorf™; Finnpipette™; Gilson™; Socorex™
  • Volume: 200 μL - 1000 μL
  • Graduated
  • Matching rack Cat.-No. 941 325
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 to 1000 μL
Volume (Metric) Dispensing 200 to 1000 μL
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Tip Style Universal
Packaging Easyload Rack
Quantity 60/Rack, 2400/Box
Graduated Yes
Material Polypropylene