Learn More
Fisherbrand™ Single-Channel Pipette
Aspirates and dispenses viscous liquids accurately and reliably. Fisherbrand™ Single-Channel Pipette include a lock counter to prevent accidental volume changes. Lightweight, ergonomic design minimizes repetitive strain injuries.
Brand: Fisherbrand™ FI8-104-00-9
This item is not returnable.
View return policy
Description
- Accurate and reliable pipetting
- Reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids
- Innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation
- Lock counter mechanism of variable volume pipettes prevents volume changes while pipetting
- Comfortable and effortless pipetting - light weight / ergonomic design for easy operation prevents hand stress. Ergonomic low plunger force minimizes RSI (repetitive strain injury)
- Pipette is fully autoclavable for sterile handling
- Easy and quick maintenance and adjustment with the included disassembly and calibration tool
- No loss of sample - blow out system allows last drop dispensing
- Meets following standards: DIN EN ISO 8655-1 | DIN EN ISO 8655-2 | DIN EN ISO 8655-6 | EU Directive 98/79/EG
Specifications
Autoclavable | |
Sterile | |
10 to 100 μL | |
0.5μL | |
2 Years |
1 | |
Manual Pipette | |
Lock counter mechanism, disassembly and calibration tool, sample bag with tips, grease | |
1 Each |