Learn More
Fisherbrand™ Plain Glass Plates with Bonded Spacers
Accessory for use with OmniPAGE mini vertical Electrophoresis systems.
Brand:  Fisherbrand™ VS10PGS0.75
 View more versions of this product
Product Code. 11867643
  • 21.40 â‚¬ / Pack of 2
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for use with OmniPAGE mini vertical Electrophoresis systems
Specifications
Specifications
Plain Glass Plate
100 x 100 mm
0.75mm bonded spacers
Product Suggestions
Product Suggestions
Videos
Videos
  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand and Fisher BioReagents Focus on electrophoresis
    SDS
    Documents
    Documents
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Fisherbrand™ Plain Glass Plates with Bonded Spacers >