missing translation for 'onlineSavingsMsg'
Learn More

Wuertt Celluloid™ Plasticized Drying Rack

Benchtop rack and tray for drying laboratory glassware. Wuertt Celluloid™ Plasticized Drying Rack is constructed of plasticized wire.

Brand:  Wuertt Celluloid™ 52810111

 View more versions of this product

Product Code. 10425591

  • 186.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes catch tray
  • Plasticized wire construction for durability
  • Angled stems
  • Looped stems for larger glassware
Specifications

Specifications

Drain tray
Items (44)
44
1 Pack
42 cm
17 cm
Benchtop Drainer
Plasticized Wire
Laboratory glassware
1 Pack
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Wuertt Celluloid™ Plasticized Drying Rack > Height: 61cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.